Learn More
Keener™ Autoclave Bag Elastic Closures
An alternative to wire tie closures
Brand:  Keener™ 01-815-8
Encompass_Preferred
Product Code. 12787057
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
An alternative to wire tie closures
  • High-tensile-strength elastic bands
  • Compatible with most autoclave bags
  • Remain intact at autoclaving temperatures
Specifications
Specifications
Autoclavable
Elastic
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Keener™ Autoclave Bag Elastic Closures