missing translation for 'onlineSavingsMsg'
Learn More
Kenpaullone, Tocris Bioscience™ Product Code.: 18005952

Kenpaullone, Tocris Bioscience™

Product Code. 18005952
10 mg
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy

Product Code. 18005952

Brand: Tocris Bioscience 1398/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GSK-3β inhibitor; also inhibits cdks

GSK-3beta inhibitor (IC50 = 0.23 μM). Also inhibits cyclin dependent kinases (cdks) in the sub micromolar range (reported IC50 values are 0.4, 0.68-7 and 0.85 μM for cdk1, cdk2 and cdk5, respectively). Generates induced pluripotent stem cells (iPSCs) from somatic cells when used in combination with reprogramming factors; can replace Klf4.
TRUSTED_SUSTAINABILITY

Specifications

CAS 142273-20-9
Molecular Formula C16H11BrN2O
Quantity 10 mg
Synonym kenpaullone, 9-bromopaullone, 9-bromo-7,12-dihydroindolo 3,2-d 1 benzazepin-6 5h-one, indolo 3,2-d 1 benzazepin-6 5h-one,9-bromo-7,12-dihydro, 9-bromo-7,12-dihydro-indolo 3,2-d 1 benzazepin-6 5h-one, 9-bromo-7,12-dihydro-5h-indolo 3,2-d 1 benzazepin-6-one, 9-bromo-7,12-dihydro-indolo-3,2-d-1 benzazepin-6 5h-one, 9-bromo-7,12-dihydrobenzo 2,3 azepino 4,5-b indol-6 5h-one, indolo 3,2-d 1 benzazepin-6 5h-one, 9-bromo-7,12-dihydro
InChI Key QQUXFYAWXPMDOE-UHFFFAOYSA-N
SMILES C1C2=C(C3=CC=CC=C3NC1=O)NC4=C2C=C(C=C4)Br
IUPAC Name 9-bromo-7,12-dihydro-5H-indolo[3,2-d][1]benzazepin-6-one
Molecular Weight (g/mol) 327.181
PubChem CID 3820
Formula Weight 327.18
Percent Purity >98%
Chemical Name or Material Kenpaullone
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kenpaullone, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.