missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Keratinocyte SFM (1X)
Contains L-glutamine.
Brand: Gibco™ 17005042
167.39 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
This item is not returnable.
View return policy
Description
Keratinocyte-SFM is supplied with prequalified human recombinant Epidermal Growth Factor 1-53 (EGF 1-53) and Bovine Pituitary Extract (BPE) in separate packaging. Components are not sold separately.Note:
A Keratinocyte-SFM Master File has been submitted to the FDA. Permission to cross-reference the Master File may be obtained by contacting Technical Services or your local Sales Representative.
Specifications
• 500 mL Keratinocyte-SFM Basal Medium; store in refrigerator (2–8°C) and protect from light • 25 mg Bovine Pituitary Extract (BPE); store in freezer (-5 to -20°C) and protect from light • 2.5 μg EGF, Human Recombinant; store in freezer (-5 to -20°C) and protect from light Complete Medium: store in refrigerator (2–8°C) and protect from light |
|
Serum-free | |
Mammalian Cell Culture | |
Kit | |
Sterile-filtered | |
Glutamine, Phenol Red | |
500 mL |
Keratinocyte Cells, Epithelial Cells | |
1 X | |
Liquid | |
Serum-free | |
Sterile-filtered | |
Sustainable packaging |
For Research Use Only. Not for use in diagnostic procedures.