missing translation for 'onlineSavingsMsg'
Learn More

Ketoconazole, Fisher BioReagents

Product Code. 10754344
Click to view available options
Quantity:
50 mg
100 mg
500 mg
1 g
Packaging:
Amber Glass
This item is not returnable. View return policy
This item is not returnable. View return policy
Ketoconazole is a cytochrome P450 inhibitor and demonstrates anti-psoriatic activity. An antifungal agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65277-42-1
Molecular Formula C26H28Cl2N4O4
Molecular Weight (g/mol) 534.452
InChI Key XMAYWYJOQHXEEK-SIULDFEJSA-N
Synonym ketoconazole
PubChem CID 76973198
IUPAC Name 2,2,2-trideuterio-1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone
SMILES CC(=O)N1CCN(CC1)C2=CC=C(C=C2)OCC3COC(O3)(CN4C=CN=C4)C5=C(C=C(C=C5)Cl)Cl

Specifications

Melting Point 148°C
Color White
Ignition Residue < 0.1%
Loss on Drying < 0.5%
Melting Range Inclusive between 148°C
Assay Percent Range 98%
Packaging Amber Glass
Quantity 100 mg
Merck Index 15, 5349
Formula Weight 531.44
Physical Form Solid
Chemical Name or Material Ketoconazole
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ketoconazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360F-May damage fertility.
  • H373-May cause damage to organs.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'