missing translation for 'onlineSavingsMsg'
Learn More
Ketoketal, 97%, Thermo Scientific™ Product Code.: 15411208

Ketoketal, 97%, Thermo Scientific™

Product Code. 15411208
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15411208

Brand: Thermo Scientific Alfa Aesar H55069.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ketoketal is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56309-94-5
Molecular Formula C14H22O3
Molecular Weight (g/mol) 238.327
MDL Number MFCD00809704
InChI Key ZNWLFTSPNBLXGL-UHFFFAOYSA-N
Synonym ketoketal, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexanone, 8-4-oxocyclohexyl-1,4-dioxaspiro 4.5 decane, 4-1,4-dioxaspiro 4.5 dec-8-yl cyclohexanone, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexan-1-one, bicyclohexane-4,4'-dione monoethylene ketal, 1,1'-bicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'-dione monoethylene ketal, cyclohexanone, 4-1,4-dioxaspiro 4.5 dec-8-ylShow MoreShow Less
PubChem CID 810913
IUPAC Name 4-(1,4-dioxaspiro[4.5]decan-8-yl)cyclohexan-1-one
SMILES C1CC(=O)CCC1C2CCC3(CC2)OCCO3

Specifications

Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 238.33
Percent Purity 97%
Chemical Name or Material Ketoketal

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberBC280501 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number5003F07X Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ketoketal, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.