missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher Apex Accessories

Product Code. 16572442 Shop All Thermo Scientific Products
Click to view available options
Product Type:
24 Combi Head
24 DW Heating Block
96 DW Head
96 DW Heating Block
96 KF Heating Block
96 PCR Head
Combi Head
Heating Block 24
Heating Block 96
Matrix Heating Block
PCR Plate Heating Block
Plate Adapter
Storage Tube Adapter
UV Lamp (Replacement)
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16572442

Brand: Thermo Scientific™ 24079930

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source.

KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality

  • User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
  • Provides versatile and high-speed purification with reproducible results
  • Easy to use interface with touch screen empowering the user to control all details of the application and purification process
  • BindIx PC Software for added convenience
  • Extensive selection of validated protocols for different type of applications
  • Cloud library for quick protocol retrieval
TRUSTED_SUSTAINABILITY

Specifications

Product Type 96 DW Head
Volume (Metric) Processing 96 KF plate 50 to 150 μL, 96 DW plate 50 to 1000 μL
Format 96 Deep-well Head
Capacity 96 Samples
For Use With (Equipment) KingFisher Apex
Magnetic Rods 96
Particle Size (Metric) ca. >1 μm
Type of Magnet Head 96 DW
Plate Type 96 KF, 96 DW
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.