missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KG-501, MedChemExpress
MedChemExpress KG-501 is a CREB inhibitor, with an IC50 of 6.89 μM.
72.90 € - 233.55 €
Specifications
Chemical Name or Material | KG-501 |
---|---|
CAS | 18228-17-6 |
Color | Light Yellow |
Purity Grade Notes | Research |
Molecular Formula | C17H13ClNO5P |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16483818
|
Medchem Express
HY-103299 |
10 mg |
102.60 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16466547
|
Medchem Express
HY-103299 |
5 mg |
72.90 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16460197
|
Medchem Express
HY-103299 |
50 mg |
233.55 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KG-501 is a CREB inhibitor, with an IC50 of 6.89 μM.Specifications
KG-501 | |
Light Yellow | |
C17H13ClNO5P | |
DMSO : 6 mg/mL (15.88 mM; Need ultrasonic and warming) | |
377.72 | |
98.08% | |
Solid |
18228-17-6 | |
Research | |
Naphthol AS-E phosphate | |
O=C(C1=C(OP(O)(O)=O)C=C2C=CC=CC2=C1)NC3=CC=C(Cl)C=C3 | |
377.72 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only