Learn More
KG-501, MedChemExpress
MedChemExpress KG-501 is a CREB inhibitor, with an IC50 of 6.89 μM.
Brand: Medchem Express HY-103299
This item is not returnable.
View return policy
Description
KG-501 is a CREB inhibitor, with an IC50 of 6.89 μM.Specifications
KG-501 | |
Light Yellow | |
C17H13ClNO5P | |
Naphthol AS-E phosphate | |
O=C(C1=C(OP(O)(O)=O)C=C2C=CC=CC2=C1)NC3=CC=C(Cl)C=C3 | |
377.72 | |
Research | |
COVID-19-immunoregulation |
18228-17-6 | |
Research | |
5 mg | |
DMSO : 6 mg/mL (15.88 mM; Need ultrasonic and warming) | |
377.72 | |
98.08% | |
Solid |
Research purposes only