Learn More
Lonza™ KGM™ Gold Keratinocyte Growth Medium SingleQuots™ Supplements and Growth Factors
Separately sold supplement and growth factor set for high and low density NHEK-Normal Human Epidermal Keratinocyte cultures.
Marke:  Lonza™ 192152
Artikelnummer. 11615090
  • 281.82 € / Each
Entdecken Sie weitere Sonderangebote
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Beschreibung
Beschreibung
  • Supplement and growth factor set sold separately from basal media.
  • Can be used with 1 bottle of 192060 (basal media).
  • Designed to support both clonal growth and high density keratinocyte proliferation.
  • Supports superior growth and morphology of keratinocytes.
  • 1 x KBM™Gold™ Basal Medium (00192151), 500 mL
  • 1 x KGM™Gold™SingleQuots™Supplement pack (00192152) containing:
    1 x Natural Cap Vial with Hydrocortisone, 0.50 mL
    1 x Natural Cap Vial with Transferrin, 0.50 mL
    1 x Amber Vial with Epinephrine, 0.25 mL
    1 x Red Cap Vial with GA-1000, 0.50 mL
    1 x Orange Cap Vial with BPE, 2.00 mL
    1 x Green Cap Vial with hEGF, 0.50 mL
    1 x Lilac Cap Vial with Insulin, 0.50 mL
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
High and low density NHEK-Normal Human Epidermal Keratinocyte cultures
0.5 mL to 2.0 mL ViaLs
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote