Learn More
Lonza™ KGM™ Keratinocyte Growth Medium SingleQuots™ Supplements and Growth Factors
Supplements and growth factors kit for use with keratinocyte basal medium.
Brand: Lonza™ CC-4131
This item is not returnable.
View return policy
Description
- Separately sold SingleQuot™ kit for supplementing basal medium.
- Part of the full KGM™ Keratinocyte Cell Growth Medium BulletKit™ (catalog number CC-3111), which consists of basal medium and supplements packaged together.
- Includes the following:
- BPE Low Protein - non specific
- HEGF
- Recombinant Human Insulin 0.5%
- Hydrocortisone Solution
- GA-1000
Specifications
BPE Low Protein - non specific, HEGF, Recombinant Human Insulin 0.5%, Hydrocortisone Solution, GA-1000 | |
Keratinocyte Cells | |
Supplements and growth factors |