missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ki 20227, Tocris Bioscience™
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
Descripción
Inhibitor of c-Fms tyrosine kinase (M-CSFR, CSF1R) (IC50 values are 2, 12, 217 and 451 nM for c-Fms, VEGFR-2, PDGFRβ and c-Kit respectively). Does not inhibit Flt3, EGFR or c-Src. Suppresses osteoclast differentiation and osteolysis in a rat bone metastasis model.
Especificaciones
Especificaciones
Quantity | 10 mg |
Formula Weight | 480.54 |
Percent Purity | >97% |
Chemical Name or Material | Ki 20227 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'