Learn More
Novus Biologicals™ Kidney Matched Tumor and Normal Tissue Lysate
Two matched 1mg/mL vials (100μg/vial) of human kidney tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.
Brand:  Novus Biologicals™ NBP2-47100
Product Code. 18120188
  • 474.00 € / Pack of 2
Explore more special offers
Description
Description
The Kidney Matched Tumor & Normal Tissue Lysate is derived from human.
Specifications
Specifications
Kidney
2 Vials
Human
Store at −80°C. Avoid freeze-thaw cycles.