Learn More
DWK Life Sciences Kimble™ Kim-Kap™ Closures
Available in natural (clear) or choice of four colors
Brand:  DWK Life Sciences 73662-16
 View more versions of this product
Product Code. 10024174
  • 160.00 € / Pack of 1000
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Available in natural (clear) or choice of four colors
  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Push Cap
Polypropylene
Non-sterile
Reusable
Autoclavable
Red
16 mm O.D. Plain-end Culture Tubes