missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ G60 Medium Duty Polyurethane-Coated Cut-Resistant Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- Constructed of ultra-high molecular weight polyethylene (UHMWPE) fibers to provide breathability and protection
- Polyurethane coated palm and fingers enhance dexterity and grip capabilities
- Separate right and left gloves in a pair provide a snug fit
Specifications
Specifications
ANSI Abrasion Rating | ANSI 4 |
Coating Material | Polyurethane |
Color | Gray |
Cut Resistance Rating | ANSI A2 |
Hand | Specific |
Latex Free | Yes |
Material | HPPE |
Product Type | Cut-resistant Gloves |
Size | Small |
EN388 Standard | EN 388 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction