missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ G80 Nitrile Chemical Resistant Gloves
Green, hand-specific gloves provide maximum protection against chemical and mechanical hazards. G80 Chemical Resistant Latex-Free Gloves include improved, more consistent cotton flock lining for added comfort.
Brand: Kimberly-Clark Professional™ 94446
37.42 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Hand specific latex-free gloves protect hands against chemical or mechanical hazards
- Diamond embossed fingertips offer enhanced wet and dry grip
- Flock-lined for easy donning and extra comfort in extended use
- Improved AQL of 0.65 versus 1.5, resulting in fewer pinholes
- Modified Nitrile formulation for more durability and enhanced chemical resistance.
- Now available in 5 bulk-packed poly bags versus individually wrapped pairs
- Ideal for: providing PPE category 3 protection, level 4 abrasion resistance and durability in manufacturing, transport, construction, agriculture cleaning and janitorial work, when handling chemicals, oils, grease, acids, caustics or solvents
Specifications
FDA, ASTM D3767, ASTM D412, ASTM D5151, ASTM D6319 | |
No | |
Yes | |
Medium | |
0.38 mm | |
No | |
JKLNOP | |
Acids, bases, Solvents and Caustics | |
Cotton Flock | |
94446 | |
13 in. |
CE 0120 | |
Green | |
Nitrile | |
15 mil | |
EN 388:4101X | |
Type A | |
Bacteria, Fungi only | |
Rough | |
Lined | |
KleenGuard™ G80 | |
33 cm |