missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ KleenGuard™ G40 Foam Nitrile Coated Multi-Purpose Gloves

Product Code. 11843480
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

KleenGuard™ G40 Foam Nitrile Coated Multi-Purpose Gloves are designed for jobs that require strength, grip, and dexterity. Seamless knit nylon shell is breathable for comfort, with a foam nitrile coating for enhanced grip capabilities.

  • Designed for jobs where strength, grip and dexterity are a must
  • Protective gloves feature oil and industrial grease grip for general purpose tasks
  • Maintain a high degree of abrasion resistance (EN 388) even after 5 wash cycles
  • 25% better productivity than leather gloves with similar capabilities
  • Seamless knit nylon on the back of the hand is breathable for a comfortable fit
  • Silicone-free

Specifications

Certifications/Compliance (Europe) EN 388
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile Foam
Color Black
Exterior Finish Partially Textured
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Medium
Style Full-hand Mitt
For Use With (Application) Automotive Assembly, Machine Repairs, and Parts Handling
Glove Cuff Style Knit
Series G40
Length (English) 9.1 in.
Length (Metric) 23 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.