missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ KleenGuard™ G40 Black Polyurethane Coated Nylon Gloves

Product Code. 15354252
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Multi-purpose protective gloves designed for tasks that require high dexterity and fine motor skills. The thin-mil design provides tactile sensitivity, making KleenGuard™ G40 Black Polyurethane Coated Nylon Gloves ideal for performing delicate tasks such as electronics assembly.

  • General purpose protective gloves designed for tasks that require high dexterity and fine motor skills
  • Thin mil design provides tactile sensitivity, ideal for performing delicate tasks
  • Seamless knit nylon backing enhances durability, and provides resistance to tears and abrasion
  • Dark color is ideal for hiding dirt

Specifications

Coated Area Palm, Fingers
Coating Material Polyurethane
Color Black
Exterior Finish Partially Textured
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size X-Large
Style Full-hand Mitt
For Use With (Application) Small Parts Handling, Precision Assembly, and Inspection
Glove Cuff Style Knit
Series G40
Length (English) 9.8 in.
Length (Metric) 25 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.