missing translation for 'onlineSavingsMsg'
Learn More
Ansell Kimtech™ A8 Non-Sterile Cleanroom Shoe Covers, Unitrax Product Code.: 15319470
Add to basket

Ansell Kimtech™ A8 Non-Sterile Cleanroom Shoe Covers, Unitrax
GREENER_CHOICE

Product Code. 15319470
Small/Medium
Click to view available options
Size:
Small/Medium
Universal
X-Large/2X-Large
This item is not returnable. View return policy

Product Code. 15319470

Brand: Ansell 39370

Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Seamless, non-skid Kimtech™ A8 Non-Sterile Cleanroom Shoe Covers are low-lint for contamination control in environments where wet splash and dry particle protection are needed.

  • 300 Kimtech™ A8 Shoe Covers / Case, 3 Bags of 100 Covers; White
  • Superior non-skid design has 4 times more grip than leading alternatives
  • Seamless bottom for better comfort; elastic ankle opening
  • Low lint for contamination control
  • Not made with natural rubber latex
  • Made with poly-film laminated fabric
  • Double bagged for use in clean manufacturing and quality control environments
  • Elastic ankle opening for a snug fit
  • Recyclable through the RightCycle Program, which turns used lab coats into raw materials for eco-friendly consumer products
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable

Specifications

Size Small/Medium
Material Spunbond, Polyethylene, Polyester
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Non-skid
Seam Type Seamless
For Use With (Application) Cleanrooms, Manufacturing
Packaging Quantity 300/Cs.
Packaging Type Double-bagged, 3 Bags of 100 Covers
Resistance None
Product Type Controlled Environments Shoe Covers
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell Kimtech™ A8 Non-Sterile Cleanroom Shoe Covers, Unitrax >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.