missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ 20mL Glass Screw-Thread Scintillation Vials

Manufactured from clear KG-33 borosilicate glass

Brand:  DWK Life Sciences 74505-20

 View more versions of this product

Product Code. 17731088

  • 287.00 € / Pack of 500
Estimated Shipment: 15-01-2026
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Manufactured from clear KG-33 borosilicate glass

Variety of cap materials and liners available.

Order Info

Vials are packaged in cellular trays having alpha-numeric coordinates for fast and easy identification. Trays are shrink wrapped to preserve cleanliness. Each tray contains 100 vials, five trays per case. Caps come in polyethylene bags of 100.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Foamed Polyethylene
Borosilicate Glass
Fast and easy identification
0.67 oz.
20 mL
Attached Screw Cap
ASTM E438
White
20 mL
Cellular Trays
Kimble
Polypropylene
100 Cases (Vial), 100 Bag (Cap)
Scintillation Vial
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.