Learn More
DWK Life Sciences Kimble™ Adapters for Round-Bottom Centrifuge Tubes
Required when centrifuging select Kimble high-strength round-bottom centrifuge tubes to 13,000xG in 50mL rotor
50.05 €
Specifications
Material Rubber
View More Specs
Products 1
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
11819913
View Documents
DWK Life Sciences
45550-15
15mL Round bottom centrifuge tube
50.05 €
Pack of 2
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
Description
Description
Required when centrifuging select Kimble high-strength round-bottom centrifuge tubes to 13,000xG in 50mL rotor
  • Made of rubber
Specifications
Specifications
Rubber