Learn More
DWK Life Sciences Kimble™ AmberScrew Thread Sample Vials with Unattached PTFE-Faced/White Rubber Lined Caps
Unattached, closed top black phenolic closures
Specifications
Material (Liner) PTFE/Rubber
Color Amber
Material Borosilicate Glass
Thread Style Screw
Closure Material Phenolic
View More Specs
Description
Description
Unattached, closed top black phenolic closures
  • Manufactured from amber 51 expansion borosilicate glass
Specifications
Specifications
PTFE/Rubber
Borosilicate Glass
Phenolic
Amber
Screw
USP Type I