Learn More
DWK Life Sciences Kimble™ AmberScrew Thread Vials w/Attached PTFE-Faced/White Rubber Lined Closures
Attached closed top black phenolic closures
682.00 €
Specifications
Material (Liner) PTFE/Rubber
Color Amber
Material Borosilicate Glass
Thread Style Screw
Closure Material Phenolic
View More Specs
Products 1
Product Code Brand Diameter (Metric) Outer Volume (Metric) Height (Metric) Volume (English) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Volume (Metric) Height (Metric) Volume (English) Price Quantity & Availability  
11833023
View Documents
DWK Life Sciences
60911B12
12 mm 1.8 mL 35 mm 0.5 dr.
682.00 €
Pack of 288
Estimated Shipment: 12-03-2025
to see stock.
Add to basket
Description
Description
  • Manufactured from amber 51 expansion borosilicate glass
  • Closures with PTFE-faced/white rubber liners or white rubber liners
  • Vials packaged in corrugated trays with partitions
Specifications
Specifications
PTFE/Rubber
Borosilicate Glass
Phenolic
USP Type I
Amber
Screw
Screw Cap (Closed-top)