missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ BioMasherII™ Closed System Micro Tissue Homogenizer
Ensure efficient grinding of samples by minimizing potential exposure to hazardous materials
223.00 € - 313.00 €
Specifications
Capacity | 0.05 oz., 1.5mL |
---|---|
Color | Clear |
Autoclavable | Autoclavable |
Capacity (Metric) | 1.5 mL |
Includes | Polypropylene tube, Polyacetal pestle |
Product Code | Brand | Packaging | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging | Sterility | Price | Quantity & Availability | ||||
13440289
|
DWK Life Sciences
749625-0010 |
Bulk | Non-sterile |
223.00 €
Pack of 50 |
Estimated Shipment: 13-03-2025
Log in to see stock. |
||||
15344182
|
DWK Life Sciences
9749625002 |
Single pack | Sterile |
313.00 €
Pack of 50 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||||
Description
Ensure efficient grinding of samples by minimizing potential exposure to hazardous materials
- Clear polypropylene tube
- Polyacetal pestle with molded-in shaft guard
- Abrasive surface (pestle tip and inner tube area)
- Available in Sterile and non-sterile versions
- Capacity: 1.5mL
For animal tissues and organs, plant materials, insects, DNA, RNA, proteins, yeasts and enzymes
Specifications
0.05 oz., 1.5mL | |
Autoclavable | |
Polypropylene tube, Polyacetal pestle | |
For animal tissues and organs, plant materials, insects, DNA, RNA, Proteins, yeasts and Enzymes |
Clear | |
1.5 mL | |
0.05 oz. |