missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached Closures

Attached open top white polypropylene closures

Specifications

Classification A
Material (Liner) PTFE/Silicone
Color Clear
Diameter (Metric) Outer 28 mm
Material Borosilicate Glass
View More Specs

Description

Description

  • Manufactured from clear 33 Expansion borosilicate glass
  • Closures supplied with PTFE-faced silicone liners
  • Vials are packaged in corrugated trays with partitions

Specifications

Specifications

A
Clear
Borosilicate Glass
Polypropylene
PTFE/Silicone
28 mm
Screw
Screw Cap (Open-top)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.