missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials with Unattached PTFE-Faced/White Rubber Lined Closures

Unattached closed top black phenolic closures

Specifications

Closure Color Black
Classification A
Material (Liner) PTFE/Rubber
Color Clear
Material Borosilicate Glass
View More Specs

Description

Description

  • Manufactured from clear 33 expansion borosilicate glass
  • Closures includes PTFE-faced/white rubber liners

Specifications

Specifications

Black
PTFE/Rubber
Borosilicate Glass
Phenolic
ASTM Type I
A
Clear
Screw
Screw Cap
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.