Learn More
DWK Life Sciences™ Kimble™ KIMAX™ Distilling Apparatus Complete Assembly
Complete assembly
749.00 €
Specifications
Type Distillation Apparatus
Material Borosilicate Glass
Certifications/Compliance USP Type I and ASTM E438, Type I, Class A
Standard Taper Joint 19/38
View More Specs
Products 1
Product Code Brand Capacity Standard Taper Stopper No. For Use With (Application) Price Quantity & Availability  
Product Code Brand Capacity Standard Taper Stopper No. For Use With (Application) Price Quantity & Availability  
12754037
View Documents
DWK Life Sciences™
21500-1000
33.81 oz., 1000mL 27 General purpose distillation and phenol distillation
749.00 €
Each
Description
Description
Use to distill small amounts of water or in APHA Method 4500-NH3 for determining nitrogen (ammonia) in purified drinking water, natural water and highly purified wastewaters (concentration, <20μg/L)
Specifications
Specifications
Distillation Apparatus
Borosilicate Glass
USP Type I and ASTM E438, Type I, Class A
19/38
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DWK Life Sciences™ Kimble™ KIMAX™ Distilling Apparatus Complete Assembly