missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly Component Parts

Components for Ultra-ware™ microfiltration assembly

137.00 €

Specifications

For Use With (Equipment) Ultra-ware microfiltration assembly with PTFE-faced fritted glass support
View More Specs

Products 1
Product Code Brand Description Material Price Quantity & Availability  
Product Code Brand Description Material Price Quantity & Availability  
11803123
View Documents
DWK Life Sciences
953753-0000
Aluminum Clamp Aluminum
137.00 €
Each
Estimated Shipment: 24-02-2026
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

Ultra-ware microfiltration assembly with PTFE-faced fritted glass support
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.