Learn More
DWK Life Sciences Kimble™ Kontes™ Dounce Tissue Grinders
Designed for cellular work in which the nucleus remains intact after homogenization
Brand:  DWK Life Sciences 885300-0000
 View more versions of this product
Product Code. 10506645
  • 335.00 € / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Designed for cellular work in which the nucleus remains intact after homogenization
All-glass construction
Order Info
Items are sold one per case. Supplied with two pestles: large (for initial sample reduction) and small (to form the final homogenate)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.0005 to 0.0025 in.
0.01 oz.
Glass
Designed for cellular work in which the nucleus remains intact after homogenization
37mm
0.06 to 0.13mm
0.5 mL
Dounce Tissue Grinder
1.4 in.
0.01 oz., 0.5mL