Learn More
DWK Life Sciences Kimble™ Kontes™ Dounce Tissue Grinders
Designed for cellular work in which the nucleus remains intact after homogenization
Brand:  DWK Life Sciences 885300-0007
 View more versions of this product
Product Code. 10749155
  • 346.00 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Designed for cellular work in which the nucleus remains intact after homogenization
All-glass construction
Order Info
Items are sold one per case. Supplied with two pestles: large (for initial sample reduction) and small (to form the final homogenate)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.0008 to 0.0022 in.
0.23 oz.
Glass
Designed for cellular work in which the nucleus remains intact after homogenization
125mm
0.071 to 0.11mm
7 mL
Dounce Tissue Grinder
4.9 in.
0.23 oz., 7mL