Learn More
DWK Life Sciences Kimble™ Kontes™ Dounce Tissue Grinders
Designed for cellular work in which the nucleus remains intact after homogenization
Brand:  DWK Life Sciences 885300-0001
 View more versions of this product
Product Code. 13406419
  • 310.00 € / Each
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Designed for cellular work in which the nucleus remains intact after homogenization
All-glass construction
Order Info
Items are sold one per case. Supplied with two pestles: large (for initial sample reduction) and small (to form the final homogenate)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.0005 to 0.0025 in.
0.03 oz.
Glass
Designed for cellular work in which the nucleus remains intact after homogenization
88mm
0.06 to 0.13mm
1 mL
Dounce Tissue Grinder
3.4 in.
0.03 oz., 1mL