Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware Three Hole Delivery Caps ETFE Nut Plug 1/4 in.
Click to view available options
Material:
PTFE
Description
- Ultra-Ware™ 3-Hole Delivery caps have 1/4 in.-28 threads top and bottom
- Features three 1/8 in. ID holes to accept 1/8 in. OD PTFE tubing
- PTFE caps are supplied with CTFE plugs and a TFE/PP o-ring
Specifications
Specifications
Material | PTFE |
No. per Case | 1 Cs. |
Product Type | Nut Plug |
For Use With (Equipment) | Seal port with flat bottom 1/4-28, 5/16-24 and M6 threads |