Learn More
DWK Life Sciences Kimble™ Kontes™ KIMAX™ Tooled Top Narrow Mouth Reservoir Bottle with Bottom Hose Outlet
Glass hose connection outlet is fused to the bottle
Specifications
For Use With (Application) Designed to store and discharge liquids via a bottom hose outlet
Includes Bottom hose outlet
Material Borosilicate Glass
Mouth Narrow
Product Type Reservoir Bottle
View More Specs
Description
Description
  • KimCote™ plastic coating for added safety in the event of breakage
  • With white enamel marking spot
  • Rubber stopper used to cap narrow mouth tooled top
  • Glass conforms to ASTM™ E 438, Type I, Class A specifications
  • Sizes 250mL and 500mL accept 5/16 in. I.D. flexible tubing
  • Sizes 1000mL and larger accept 3/8 in. I.D. tubing
Specifications
Specifications
Designed to store and discharge liquids via a bottom hose outlet
Borosilicate Glass
Reservoir Bottle
Bottom hose outlet
Narrow