Learn More
DWK Life Sciences Kimble™ Kontes™ Microscale B - Connector
Unique design adapts GPI threaded glass to a serrated PTFE hose connection or 0.25 in. -28 tubing connections
Brand:  DWK Life Sciences 747185-0013
Product Code. 17760878
  • 23.40 € / Each
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Fits hose I.D.: 0.25 in.
  • Internal Thread Size: 0.25 in.-28
  • GPI Thread Size: 13-425
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hose Connector
PTFE
13-425