Learn More
DWK Life Sciences Kimble™ Kontes™ Replacement Tube For All-Glass Dounce Tissue Grinders
A range of replacement tubes for Dounce Tissue Grinders, from 2mL to 100mL.
Brand:  DWK Life Sciences 885303-0015
 View more versions of this product
Product Code. 11816271
  • 131.00 € / Each
Estimated Shipment: 31-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
A range of replacement tubes for Dounce Tissue Grinders, from 2mL to 100mL.
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Case of 1
15 mL
Dounce all Glass tissue grinders
0.5 oz.
Tissue Grinder Tube
0.5 oz., 15mL