Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Solvent Filtration Solvent Pick-up Adapter
Converts Ultra-Ware filtration assemblies for inline filtration/degassing applications
Brand: DWK Life Sciences 953906-0047
334.85 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
- Replace funnels with this adapter to support base in the same manner that funnels do
Specifications
PTFE |