missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached White Rubber Lined Caps

Packaged in corrugated trays with partitions

Specifications

Classification A
Material (Liner) Rubber
Color Clear
Material Borosilicate Glass
Thread Style Screw
View More Specs

Products 11
Product Code Brand Volume (Metric) Diameter (Metric) Outer Height (Metric) Quantity Price Quantity & Availability  
Product Code Brand Volume (Metric) Diameter (Metric) Outer Height (Metric) Quantity Price Quantity & Availability  
11812953
View Documents Promotion Details
DWK Life Sciences
60811D-10
40 mL 28 mm 95 mm 72/Cs. N/A
N/A
11842973
View Documents Promotion Details
DWK Life Sciences
60811D-6
24 mL 23 mm 85 mm 144/Cs. N/A
N/A
11802983
View Documents Promotion Details
DWK Life Sciences
60811D1738
4 mL 17 mm 38 mm 200/Cs. N/A
N/A
11802973
View Documents Promotion Details
DWK Life Sciences
60811D-2
8 mL 17 mm 60 mm 144/Cs. N/A
N/A
11832973
View Documents Promotion Details
DWK Life Sciences
60811D4
16 mL 21 mm 70 mm 144/Cs. N/A
N/A
11823023
View Documents Promotion Details
DWK Life Sciences
60811D-12
2 mL 12 mm 35 mm 288/Cs. N/A
N/A
11892973
View Documents Promotion Details
DWK Life Sciences
60811D-1528
2 mL 15 mm 28 mm 200/Cs. N/A
N/A
11812983
View Documents Promotion Details
DWK Life Sciences
60811D1940
6 mL 19 mm 40 mm 200/Cs. N/A
N/A
11822953
View Documents Promotion Details
DWK Life Sciences
60811D5
20 mL 28 mm 57 mm 72/Cs. N/A
N/A
11812973
View Documents Promotion Details
DWK Life Sciences
60811D-3
12 mL 19 mm 65 mm 144/Cs. N/A
N/A
11892963
View Documents Promotion Details
DWK Life Sciences
60811D-11
4 mL 15 mm 45 mm 144/Cs. N/A
N/A
Description

Description

  • Small case quantities
  • Manufactured from clear 33 expansion borosilicate glass
  • Attached closed top black phenolic closures

Specifications

Specifications

A
Clear
Screw
Screw Cap (Closed-top)
Rubber
Borosilicate Glass
Phenolic
Videos
SDS
Documents

Documents

Product Certifications