Learn More
DWK Life Sciences Kimble™ Closures for Sample Vials
Black phenolic closure with PTFE-lined cap
154.00 € - 383.00 €
Specifications
Product Type Screw Cap
Liner Material PTFE, Rubber
Material Phenolic
Color Black
View More Specs
Products 4
Product Code Brand Closure Size Price Quantity & Availability  
Product Code Brand Closure Size Price Quantity & Availability  
17781078
View Documents
DWK Life Sciences
73802-18400
18-400
154.00 €
Pack of 144
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
15365536
View Documents
DWK Life Sciences
73802-15425
15-425
224.00 €
Pack of 144
Estimated Shipment: 18-06-2025
to see stock.
Add to basket
10701574
View Documents
DWK Life Sciences
73802-20400
20-400
383.00 €
Pack of 144
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
11853133
View Documents
DWK Life Sciences
73802-24400
24-400
289.00 €
Pack of 144
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
Description
Description
Black phenolic closure with PTFE-lined cap
  • Works with media bottles and screw-thread vials
  • Special black phenolic material (wood filled) very resistant to the effects of temperature and steam encountered in autoclaving
  • Cemented-in PTFE-faced rubber liner is provided
Specifications
Specifications
Screw Cap
PTFE, Rubber
Phenolic
Black