Learn More
DWK Life Sciences Kimble™ Glass Screw-Thread Sample Vials without Closure
Cylindrical shape provides solid balance. Flat bottom facilitates free-standing capability.
Brand:  DWK Life Sciences 60810-2170
 View more versions of this product
Product Code. 17741078
  • 441.00 € / Pack of 1000
Estimated Shipment: 17-07-2026
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Glass sample vial has a screw thread finish
  • KG-33 borosilicate glass
  • Ungraduated with no closure included
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Borosilicate Glass
Screw
Screw Cap
ASTM Type I, Class A
21 mm
16.0 mL
1 dr.
70 mm
18-400