Learn More
DWK Life Sciences Kimble™ KIMAX™ 5-Place NMR Tube Washers
These complete five-place washers use flexible PTFE tubing and a vacuum source (not included) to direct a power stream of wash solvent to thoroughly clean the insides of 7 and 8 in. NMR tubes.
928.00 €
Specifications
Material PTFE or Borosilicate Glass
Product Type NMR Tube Washer
View More Specs
Products 1
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
14820422
View Documents
DWK Life Sciences
897033-0005
5 mm dia. Tubes
928.00 €
Each
Estimated Shipment: 01-01-2026
to see stock.
Add to basket
Description
Description
These complete five-place washers use flexible PTFE tubing and a vacuum source (not included) to direct a power stream of wash solvent to thoroughly clean the insides of 7 and 8 in. NMR tubes.
  • Two sizes available for 3 and 5mm diameter tubes
  • Both sizes come with a 1L reservoir bottle
  • All wetted parts are constructed of chemical-resistant PTFE or borosilicate glass
  • An O-ring slips over each tube to form a vacuum-tight seal before tubes are positioned for cleaning
  • With a simple twist, the wash valve can be configured to wash and then dry
  • Cycles can be repeated, or cleaned tubes can be removed immediately for use
Specifications
Specifications
PTFE or Borosilicate Glass
NMR Tube Washer
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DWK Life Sciences Kimble™ KIMAX™ 5-Place NMR Tube Washers