missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ KIMAX™-51 Brand Reusable Class B Mohr (Measuring) Pipets

Rugged construction

Specifications

Certifications/Compliance ASTM E438, USP Type I, Class B
Class B
Material Borosilicate Glass
Graduated Yes
Type Graduated Pipet, Type 1
View More Specs

Products 2
Product Code Brand Volume (Metric) Color Tolerance Price Quantity & Availability  
Product Code Brand Volume (Metric) Color Tolerance Price Quantity & Availability  
12645866
View Documents Promotion Details
DWK Life Sciences
37020-10
10 mL Orange ±0.06 mL N/A
N/A
15252676
View Documents Promotion Details
DWK Life Sciences
37020-110
0.1 mL White ±0.005 mL N/A
N/A
Description

Description

Rugged construction

  • Intended for chemical laboratory work—tip openings are smaller than usually desired for clinical laboratory operations
  • Permanent brown, graduated scale fused into uniform bore tubing ending above taper
  • Color coded by size for ease in sorting
  • Tempered tips
  • Calibrated “To Deliver”

Compliance

Designed from ASTM™ E 1293, Class B, Style 1 requirements.

Specifications

Specifications

ASTM E438, USP Type I, Class B
Borosilicate Glass
Graduated Pipet, Type 1
B
Yes
Reusable
Videos
SDS
Documents

Documents

Product Certifications