missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Pellet Pestle™ Microtubes

For use with Pellet Pestle

Brand:  DWK Life Sciences 95199119

 View more versions of this product

Product Code. 13111074

  • 62.25 € / Pack of 100
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For use with Pellet Pestle

  • Choose autoclavable polypropylene or RNase-, DNase-, pyrogen-free material (manufactured under M5.5 [Class 10,000] cleanroom conditions)
  • Individually wrapped

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.05 oz.
RNase-, DNase-, pyrogen-free
Yes
1.5 mL
Microtube
0.05 oz., 1.5mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers