missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Reagent Sprayers with Screw Thread Ground Joint

Product Code. p-73088
Click to view available options
Capacity (Metric):
50 mL
Diameter (Metric) Outer:
5 cm
Standard Taper Joint:
19/22
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11773143

Brand: DWK Life Sciences 4225300050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provide uniform spray mist of visualization reagents for TLC and paper chromatography; use with all standard TLC reagents and most corrosive liquids

  • Liquid touches only nonreactive borosilicate glass atomizing sprayer head and flask
  • Sprayer head-to-flask connection consists of screw-thread standard taper ground joint, connecting cap, and loosening ring
  • Operate sprayer at 3 to 5psi (0.021 to 0.035MPa)
  • Adjust spray pattern by varying the position of thumb on vent hole
  • Autoclavable (after cap, loosening ring, and O-ring are removed)

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Capacity (English) 1.69 oz.
Capacity (Metric) 50 mL
Connections Screw thread ground joint, connecting cap, loosening ring
Material Glass
Quantity 1 Case
Type TLC Reagent Sprayer
For Use With (Application) To Provide Uniform Spray Mist of Visualization Reagents for TLC and Paper Chromatography
For Use With (Equipment) Standard TLC Reagents and Most Corrosive Liquids
Operating Pressure 3 to 5 psi
Standard Taper Joint 19/22
Height (English) 6.85 in.
Height (Metric) 17.4 cm
Diameter (English) Outer 1.96 in.
Diameter (Metric) Outer 5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.