missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Phenolic Caps with Pulp/Vinyl Liners

Product Code. 15384052
Click to view available options
Neck Finish:
20-400
This item is not returnable. View return policy
Not recommended for hydrocarbons or bleaches.
This item is not returnable. View return policy
Not recommended for hydrocarbons or bleaches.

Cap provides good chemical resistance to mild acids, alkalis, alcohols, aqueous solutions, oils and solvents.

  • Polyvinyl film is adhered to 0.035-in. pulpboard
  • Color: black
  • Low cost cap for general purpose usage
  • Not recommended for hydrocarbons or bleaches

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 144 Cs.
Neck Finish 20-400
Product Type Phenolic Cap
For Use With (Equipment) Bottle
Closure Liner Pulp/Vinyl
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'