missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Score-Break Trimmed-Stem Ampules

Offer maximum solution stability, easy sealing and opening

Brand:  DWK Life Sciences 12040G-2

 View more versions of this product

Product Code. 17760858

  • 74.35 € / Pack of 144
Estimated Shipment: 15-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Choice of Amber 203 or Kimax-51™ clear
  • United States Pharmacopeia (USP) Type I borosilicate glass
  • Pull sealed
  • Score-break, trimmed stem for easy opening
  • Special ampule file not required

Amber ampules

  • 1mL and 2mL
  • Kimax-51™ ampules
    • 2mL and 20mL

    TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    2 mL
    Prescored Ampule
    12 mm
    7.5cm
    3cm
    Borosilicate Glass
    Amber
    0.47 in.
    2.95 in.
    1.18 in.
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.