Learn More
DWK Life Sciences Kimble™ Stoppers for Headspace Vials
Gray stoppers for aluminum seal finish
Brand:  DWK Life Sciences 73827-21
Product Code. 17711088
  • 328.00 € / Pack of 1000
Estimated Shipment: 16-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Butyl rubber
  • Fits GPI aluminum seal finish 20 mm
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gray
Butyl
Aluminum Seal
20