missing translation for 'onlineSavingsMsg'
Learn More
Ansell Kimtech™ A5 Sterile Integrated Hood and Mask Product Code.: 15190077
In Stock
to see stock.
Add to basket

Ansell Kimtech™ A5 Sterile Integrated Hood and Mask
GREENER_CHOICE

Product Code. 15190077
Universal
Click to view available options
Size:
Universal
This item is not returnable. View return policy

Product Code. 15190077

Brand: Ansell 36072

In Stock
to see stock.
Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Cleanroom sterile hood with a mask integrated into the hood.

  • Average bacterial filtration efficiency (BFE) of 96%
  • Polypropylene breathable SMS fabric
  • Triple-bagged and vacuum packaged
  • Suitable for ISO Class 5 or higher cleanrooms
  • Clean-don ties for easy gowning and are folded inside out for aseptic donning
  • Protective hood-and-mask-combos are made without latex, silicone or the BHT preservative
  • Designed for single-use
  • CE certified, Category III
  • Each individually bagged mask and hood has a sterility indicator
  • Manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable

Specifications

Product Type Integrated Hood and Mask
Size Universal
Material Polypropylene SMS Fabric
Color White
Closure Type Ties
Disposable Single-use
Latex Free Yes
Model 36072
Sterility Sterile
Certifications/Compliance Type 6, Category III PPE
Quantity 75/Cs.
Includes Pull down ties, stretch-fit elastic, blue donning line
For Use With (Application) Laboratory Protection
Hood Type Integrated
Cleanroom Class Class 100 (ISO 5)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell Kimtech™ A5 Sterile Integrated Hood and Mask >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.