missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ Kimtech™ Centrefeed Wipers

Centrefeed, white wipers for wiping jobs where precision and reliability are essential. Kimtech™ Centrefeed Wipers for excellent absorbency and low-lint wiping. Acid, base and solvent resistant.

Brand:  Kimberly-Clark Professional™ 61930

193.80 EUR valid until 2025-02-13
Use promo code "24130" to get your promotional price.


Product Code. 17112242

  • 333.00 € / 6 rolls
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Engineered for cloth-like softness, excellent absorbency and low-lint wiping with low extractables (no binders or surfactants)
  • Designed to work well with solvents, acid, base and solvent-resistant)
  • Delivered in a centrefeed system, quick loading, easy-clean dispenser Ideal for any environment requiring fast, easy and hygienic access to wipers
  • Uses:
    • For a specific task in controlled areas like transport, healthcare and manufacturing
    • Ideal for wiping jobs where precision and reliability are essential such as clean manufacturing and printing environments
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Heavy-duty Wiper
    48.6 cm
    17.8 cm
    White
    Suited to Specific Tasks in Controlled Areas Like Transport, Healthcare and Manufacturing
    Non-sterile
    19.1 in.
    7 in.
    Virgin Wood Fiber
    6 Rolls x 225 Sheets, White (1,350 total)
    6 Rolls x 225 Sheets (1,350 total)
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.