missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Kimtech™ Polishing Cloths Bucket

Designed for dry polishing processes. Kimberly-Clark™ Kimtech™ Polishing Cloths Bucket contains 300 spunlace, white cloths suitable for surface preparation, pre/post paint applications, and polishing glass.

Brand:  Kimberly-Clark™ 7213

182.09 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 12695586

  • 497.00 € / Pack of 300
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with the fabricated metal manufacturing industry
  • 300 white sheets on a roll, dispensed from a specially designed bucket style dispenser
  • Removable lid for convenient refilling
  • Separate centrefeed lid for easy dispensing of sheets and controlled usage.
  • Durable metal handle and plastic hand grip
  • Ideal for wiping on the move
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polishing Cloth Bucket Dispenser
60 x 39 cm
60 cm
Paint applications and for polishing Glass surfaces
White
14.82 in.
39 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.