Learn More
Kimberly-Clark™ Kimtech™ Pure W4 Wipers
Use these cloths for dry wiping of critical equipment and surfaces in pharmaceutical, high-tech biotechnology, medical device industries and food processing industries. Kimtech™ W4 Wipers are suitable for ISO Class 4 or higher cleanroom environments. Double bagged with case liner.
358.00 €
Specifications
Product Type Cleanroom Wiper
Color White
Sterility Non-sterile
View More Specs
Products 1
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
12685606
View Documents
Kimberly-Clark™
7605
30.4 x 30.4 cm
358.00 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
Description
Description
Wipers are thermally bonded with no glue or binding and straight, clean, knife-cut edges to minimize lint.
  • Low linting, 100% Polypropylene white base sheets
  • Acid, base and solvent resistant
  • Ideal for pharmaceutical, high-tech biotechnology, medical device and food processing industries
  • Suitable for ISO Class 4, or higher, clean-room environments
  • 100 Sheets
  • Doubled-bagged with case liner
Specifications
Specifications
Cleanroom Wiper
White
Non-sterile