missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Kimtech Science™ Delicate Task Wipes
GREENER_CHOICE

Provides high-performance cleaning in the laboratory. Kimtech Science™ Delicate Task Wipes are suitable for ISO Class 4 or higher laboratory environments.

Brand:  Kimberly-Clark™ 7558

70.50 EUR valid until 2024-12-31
Use promo code "24046" to get your promotional price.


Product Code. 15402680

  • 132.00 € / Pack of 15
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Ideal for cleaning of delicate glassware, instruments and surface/parts
  • With multi-ply absorbency and low lint content for intricate dusting
  • Dispensed from a POP-UP Box flexible system, for hygiene and controlled access
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Light-duty Industrial Wiper
20.5 cm
20 cm
Single-use
15 x 200 Sheets
Non-sterile
8.3 in.
8.3 in.
Paper
20.5 x 20 cm
Multi Ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.